missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSP50 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2430
This item is not returnable.
View return policy
Description
TSP50 Monoclonal specifically detects TSP50 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TSP50 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 29122 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| 260606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PRSS50 | |
| Mouse myeloma cell line NS0-derived recombinant human TSP50 Ala40-Ala363 (predicted) | |
| 100 μg | |
| Primary | |
| Detects human TSP50 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactvity with recombinant mouse TSP50 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction