missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Troponin I/TNNI3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£168.00 - £379.00
Specifications
| Antigen | Troponin I Type 3 (cardiac) |
|---|---|
| Clone | 923952 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159730
|
R&D Systems
MAB85942-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109080
|
R&D Systems
MAB85942-100 |
100 μg |
£379.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for ELISA.Specifications
| Troponin I Type 3 (cardiac) | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1, TNNC1troponin I, cardiac, TNNI3,, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
| E. coli-derived recombinant human Troponin I/TNNI3, Met1-Ser210 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Troponin I/TNNI3 in direct ELISAs. |
| 923952 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 7137 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title