missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Troponin I/TNNI3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB85942-100
This item is not returnable.
View return policy
Description
Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for ELISA.
Specifications
| Troponin I Type 3 (cardiac) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Troponin I/TNNI3 in direct ELISAs. | |
| Human | |
| Supernatant |
| ELISA | |
| 923952 | |
| ELISA | |
| Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1, TNNC1troponin I, cardiac, TNNI3,, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
| E. coli-derived recombinant human Troponin I/TNNI3, Met1-Ser210 | |
| 100 μg | |
| 7137 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction