missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREM2 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
£189.00 - £412.00
Specifications
| Antigen | TREM2 |
|---|---|
| Clone | 2958E |
| Dilution | Immunohistochemistry 1-10 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279903
|
R&D Systems
MAB11618-SP |
25 μg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279926
|
R&D Systems
MAB11618-100 |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for ImmunohistochemistrySpecifications
| TREM2 | |
| Immunohistochemistry 1-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse myeloma cell line, NS0-derived human TREM2, His19-Ser174, Accession # Q9NZC2 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 2958E | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9NZC2 | |
| 54209 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title