missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREM2 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11618-SP
This item is not returnable.
View return policy
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for Immunohistochemistry
Specifications
| TREM2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse myeloma cell line, NS0-derived human TREM2, His19-Ser174, Accession # Q9NZC2 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Immunohistochemistry | |
| 2958E | |
| Immunohistochemistry 1-10 μg/mL | |
| Q9NZC2 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54209 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction