missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREM2 Antibody, R&D Systems™

Goat nan Antibody
£193.00 - £516.00
Specifications
| Antigen | TREM2 |
|---|---|
| Dilution | ELISA |
| Applications | ELISA |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779626
![]() |
R&D Systems
AF18281-SP |
25 μg |
£193.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788936
![]() |
R&D Systems
AF18281 |
100 μg |
£516.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TREM2 nan antibody specifically detects TREM2 in Human samples. It is validated for ELISASpecifications
| TREM2 | |
| ELISA | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse myeloma cell line NS0-derived recombinant human TREM2, His19-Ser174, Accession # Q9NZC2 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. |
| ELISA | |
| Unconjugated | |
| Goat | |
| Human | |
| Q9NZC2 | |
| 54209 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title