missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREM2 Antibody, R&D Systems™

Goat nan Antibody
Brand: R&D Systems AF18281
This item is not returnable.
View return policy
Description
TREM2 nan antibody specifically detects TREM2 in Human samples. It is validated for ELISA
Specifications
| TREM2 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse myeloma cell line NS0-derived recombinant human TREM2, His19-Ser174, Accession # Q9NZC2 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| ELISA | |
| ELISA | |
| Q9NZC2 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 54209 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction