missing translation for 'onlineSavingsMsg'
Learn More

Human TREM-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 13 publications

£181.00 - £1895.00

Specifications

Antigen TREM1
Clone 193015
Concentration LYOPH
Dilution Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity, CyTOF-ready
Applications Flow Cytometry, CyTOF
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18754317
GREENER_CHOICE
View Documents
R&D Systems
MAB1278-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18771458
GREENER_CHOICE
View Documents
R&D Systems
MAB1278-100
100 μg
£467.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18702448
GREENER_CHOICE
View Documents
R&D Systems
MAB1278-500
500 μg
£1895.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF-ready.
Specifications

Specifications

TREM1
LYOPH
Flow Cytometry, CyTOF
Unconjugated
Mouse
Agonist Activity
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
TREM1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
193015
Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity, CyTOF-ready
Monoclonal
Purified
RUO
Human
Q9NP99
54210
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Ala21-Asn205 Accession # Q9NP99
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human TREM-1 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.