Learn More
Human TREM-1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 13 publications
Brand:  R&D Systems MAB1278-SP
 View more versions of this product
Product Code. 18754317
  • £181.00 / 25µg
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TREM1
Monoclonal
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity, CyTOF-ready
Q9NP99
TREM1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Ala21-Asn205 Accession # Q9NP99
25 μg
Agonist Activity
54210
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Flow Cytometry, CyTOF
193015
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects human TREM-1 in direct ELISAs.
Human
Purified