missing translation for 'onlineSavingsMsg'
Learn More

Human Trappin-2/Elafin Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£156.00 - £497.00

Specifications

Antigen Trappin-2/Elafin/Skalp
Clone 257729
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18745529
GREENER_CHOICE
View Documents
R&D Systems
MAB1747-SP
25 μg
£156.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18162421
View Documents
R&D Systems
MAB1747
500 μg
£497.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trappin-2/Elafin/Skalp Monoclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for Western Blot.
Specifications

Specifications

Trappin-2/Elafin/Skalp
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
cementoin, Elafin, Elastase-specific inhibitor, ESI, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI3, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALP, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, Trappin2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin
PI3
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
257729
Western Blot
Unconjugated
Mouse
Human
P19957
5266
Mouse myeloma cell line NS0-derived recombinant human Trappin-2/Elafin Ala23-Gln117 Accession # P19957
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects the full length form (amino acids 23-117) and the trypsin-processed form (amino acids 61-117) of human Trappin-2/Elafin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human SLPI is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.