Learn More
Human Trappin-2/Elafin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB1747
 View more versions of this product
Product Code. 18162421
  • £497.00 / 500µg
Description
Trappin-2/Elafin/Skalp Monoclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Trappin-2/Elafin/Skalp
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
cementoin, Elafin, Elastase-specific inhibitor, ESI, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI3, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALP, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, Trappin2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5266
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot
257729
Western Blot 1 ug/mL
P19957
PI3
Mouse myeloma cell line NS0-derived recombinant human Trappin-2/Elafin Ala23-Gln117 Accession # P19957
500 μg
Primary
Detects the full length form (amino acids 23-117) and the trypsin-processed form (amino acids 61-117) of human Trappin-2/Elafin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human SLPI is observed.
Human
Purified