missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMEFF1/Tomoregulin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £442.00
Specifications
| Antigen | TMEFF1/Tomoregulin-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776127
![]() |
R&D Systems
AF1919-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18090992
![]() |
R&D Systems
AF1919 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEFF1/Tomoregulin-1 Polyclonal specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Western Blot.Specifications
| TMEFF1/Tomoregulin-1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| C9orf2, H7365member 1, Tomoregulin1, Tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1 | |
| TMEFF1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8IYR6 | |
| 8577 | |
| Mouse myeloma cell line NS0-derived recombinant human TMEFF1/Tomoregulin-1 Ser40-Thr328 Accession # Q8IYR6 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human TMEFF1/Tomoregulin-1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse TMEFF1 is observed and less than 1% cross-reactivity with recombinant human TMEFF2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title