missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMEFF1/Tomoregulin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1919-SP
This item is not returnable.
View return policy
Description
TMEFF1/Tomoregulin-1 Polyclonal specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Western Blot.
Specifications
| TMEFF1/Tomoregulin-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C9orf2, H7365member 1, Tomoregulin1, Tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8577 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q8IYR6 | |
| TMEFF1 | |
| Mouse myeloma cell line NS0-derived recombinant human TMEFF1/Tomoregulin-1 Ser40-Thr328 Accession # Q8IYR6 | |
| 25 μg | |
| Primary | |
| Detects human TMEFF1/Tomoregulin-1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse TMEFF1 is observed and less than 1% cross-reactivity with recombinant human TMEFF2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction