missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
£187.00 - £517.00
Specifications
| Antigen | TIM-3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, CyTOF-ready, Knockout Validated 1 ug/mL, Dual RNAscope ISH-IHC 3-25 ug/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, Immunoassay, In Situ Hybridization (ISH) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746077
![]() |
R&D Systems
AF2365-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028591
|
R&D Systems
AF2365 |
100 μg |
£517.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-3 Polyclonal specifically detects TIM-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, Knockout Validated, Dual RNAscope ISH-IHC.Specifications
| TIM-3 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, CyTOF-ready, Knockout Validated 1 ug/mL, Dual RNAscope ISH-IHC 3-25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
| HAVCR2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, Immunoassay, In Situ Hybridization (ISH) | |
| Unconjugated | |
| RUO | |
| Q8TDQ0 | |
| 84868 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-3 Ser22-Arg200 Accession # Q8TDQ0 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human TIM-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) TIM-3 is observed and less than 1% cross-reactivity with recombinant human TIM-1, rmTIM-1, and rmTIM-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title