missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 16 publications
Brand: R&D Systems AF2365
This item is not returnable.
View return policy
Description
TIM-3 Polyclonal specifically detects TIM-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, Knockout Validated, Dual RNAscope ISH-IHC.
Specifications
| TIM-3 | |
| Polyclonal | |
| Unconjugated | |
| Q8TDQ0 | |
| HAVCR2 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-3 Ser22-Arg200 Accession # Q8TDQ0 | |
| 100 μg | |
| Primary | |
| Detects human TIM-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) TIM-3 is observed and less than 1% cross-reactivity with recombinant human TIM-1, rmTIM-1, and rmTIM-2 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, Immunoassay, In Situ Hybridization (ISH) | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, CyTOF-ready, Knockout Validated 1 ug/mL, Dual RNAscope ISH-IHC 3-25 ug/mL | |
| CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84868 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction