missing translation for 'onlineSavingsMsg'
Learn More

Human TIM-3 Antibody, Novus Biologicals Biologicals™

Rabbit Monoclonal Antibody

£206.00 - £476.00

Specifications

Antigen TIM-3
Clone 2321B
Dilution Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL
Applications Multiplex Immunofluorescence, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306098
View Documents Promotion Details
R&D Systems
MAB11672-SP
25 μg
£206.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306121
View Documents Promotion Details
R&D Systems
MAB11672-100
100 μg
£476.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
Specifications

Specifications

TIM-3
Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3
Mouse myeloma cell line, NS0-derived recombinant human TIM-3, Met1-Arg200, Accession # Q8TDQ0
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Antibody
2321B
Multiplex Immunofluorescence, Immunohistochemistry
Unconjugated
Rabbit
Human
Q8TDQ0
84868
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.