missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-3 Antibody, Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11672-SP
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
Specifications
| TIM-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
| Mouse myeloma cell line, NS0-derived recombinant human TIM-3, Met1-Arg200, Accession # Q8TDQ0 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Multiplex Immunofluorescence, Immunohistochemistry | |
| 2321B | |
| Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL | |
| Q8TDQ0 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 84868 | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction