missing translation for 'onlineSavingsMsg'
Learn More

Human Thrombopoietin/Tpo Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£168.00 - £1570.00

Specifications

Antigen Thrombopoietin/Tpo
Clone 34817
Dilution Neutralization 5-10 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Neutralization, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18758309
GREENER_CHOICE
View Documents
R&D Systems
MAB288-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18737559
GREENER_CHOICE
View Documents
R&D Systems
MAB288-100
100 μg
£389.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18757749
GREENER_CHOICE
View Documents
R&D Systems
MAB288-500
500 μg
£1570.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Thrombopoietin/Tpo
Neutralization 5-10 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P40225
7066
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo Ser22-Gly353 Accession # P40225
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Thrombopoietin/Tpo in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) G-CSF, rhG-CSF R, rhGM-CSF, recombinant mouse GM-CSF, or rhM-CSF is observed.
34817
Neutralization, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
THPO
IgG2b
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.