missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thrombopoietin/Tpo Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB288-100
This item is not returnable.
View return policy
Description
Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Thrombopoietin/Tpo | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7066 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization, ELISA | |
| 34817 | |
| Neutralization 5-10 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P40225 | |
| THPO | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo Ser22-Gly353 Accession # P40225 | |
| 100 μg | |
| Primary | |
| Detects human Thrombopoietin/Tpo in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) G-CSF, rhG-CSF R, rhGM-CSF, recombinant mouse GM-CSF, or rhM-CSF is observed. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction