missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFF2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
£336.92
Specifications
Antigen | TFF2 |
---|---|
Clone | 366508 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Description
TFF2 Monoclonal specifically detects TFF2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TFF2 | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q03403 | |
7032 | |
E. coli-derived recombinant human TFF2 Glu24-Tyr129 Accession # Q03403 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TFF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human TFF3. |
366508 | |
Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SML1, SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, Spasmolysin, Spasmolytic polypeptide, trefoil factor 2 | |
TFF2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only