missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4077-SP
This item is not returnable.
View return policy
Description
TFF2 Monoclonal specifically detects TFF2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TFF2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL | |
| Q03403 | |
| TFF2 | |
| E. coli-derived recombinant human TFF2 Glu24-Tyr129 Accession # Q03403 | |
| 25 μg | |
| Primary | |
| Detects human TFF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human TFF3. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 366508 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SML1, SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, Spasmolysin, Spasmolytic polypeptide, trefoil factor 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7032 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction