missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACI/TNFRSF13B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£171.00 - £1765.00
Specifications
| Antigen | TACI/TNFRSF13B/CVID |
|---|---|
| Clone | 165609 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752757
![]() |
R&D Systems
MAB174-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701027
![]() |
R&D Systems
MAB174-100 |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18764417
![]() |
R&D Systems
MAB174-500 |
500 μg |
£1765.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).Specifications
| TACI/TNFRSF13B/CVID | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| O14836 | |
| 23495 | |
| Mouse myeloma cell line NS0-derived recombinant human TACI/TNFRSF13B Ser2-Thr166 Accession # O14836 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TACI/TNFRSF13B in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TACI or recombinant human BCMA is observed. |
| 165609 | |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF13B, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
| TNFRSF13B | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title