missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACI/TNFRSF13B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB174-100
This item is not returnable.
View return policy
Description
TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).
Specifications
| TACI/TNFRSF13B/CVID | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF13B, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23495 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, ELISA | |
| 165609 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O14836 | |
| TNFRSF13B | |
| Mouse myeloma cell line NS0-derived recombinant human TACI/TNFRSF13B Ser2-Thr166 Accession # O14836 | |
| 100 μg | |
| Primary | |
| Detects human TACI/TNFRSF13B in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TACI or recombinant human BCMA is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction