missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Syndecan-2/CD362 Antibody, R&D Systems™

Rat Monoclonal Antibody
£206.00 - £1730.00
Specifications
| Antigen | Syndecan-2/CD362 |
|---|---|
| Clone | 305515R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18763378
![]() |
R&D Systems
MAB2965R-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729458
![]() |
R&D Systems
MAB2965R-100 |
100 μg |
£438.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149640
|
R&D Systems
MAB2965R-01M |
1 mg |
£1460.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18171661
|
R&D Systems
MAB2965R-500 |
500 ug |
£1730.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syndecan-2/CD362 Monoclonal specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry.Specifications
| Syndecan-2/CD362 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| AAH49836 | |
| 6383 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Detects human Syndecan-2/CD362 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Syndecan-1, recombinant human (rh) Syndecan-3, or rhSyndecan-4 is observed. |
| 305515R | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Human | |
| CD362, CD362 antigen, Fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1, HSPG1syndecan proteoglycan 2, SDC2, SYND2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2, Syndecan2 | |
| SDC2 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title