missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Syndecan-2/CD362 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2965R-01M
This item is not returnable.
View return policy
Description
Syndecan-2/CD362 Monoclonal specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry.
Specifications
| Syndecan-2/CD362 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| CD362, CD362 antigen, Fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1, HSPG1syndecan proteoglycan 2, SDC2, SYND2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2, Syndecan2 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6383 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 305515R | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| AAH49836 | |
| SDC2 | |
| Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362 Glu19-Gly144 Accession No. AAH49836 | |
| 1 mg | |
| Primary | |
| Detects human Syndecan-2/CD362 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Syndecan-1, recombinant human (rh) Syndecan-3, or rhSyndecan-4 is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction