missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SVEP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £378.00
Specifications
| Antigen | SVEP1 |
|---|---|
| Clone | 987420 |
| Dilution | Western Blot 2μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18183071
![]() |
R&D Systems
MAB97741-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141311
|
R&D Systems
MAB97741-100 |
100 μg |
£378.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SVEP1 Monoclonal specifically detects SVEP1 in Human samples. It is validated for Western Blot.Specifications
| SVEP1 | |
| Western Blot 2μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q4LDE5 | |
| 79987 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 987420 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| C9orf13, CCP module-containing protein 22, CCP22, FLJ16013, FLJ90719, POLYDOM, selectin-like protein, SELOB, SEL-OB, sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1, sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1 | |
| Synthetic peptide containing human SVEP1 Peptide Accession # Q4LDE5 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human SVEP1 in direct Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title