missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SVEP1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB97741-100
This item is not returnable.
View return policy
Description
SVEP1 Monoclonal specifically detects SVEP1 in Human samples. It is validated for Western Blot.
Specifications
| SVEP1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| C9orf13, CCP module-containing protein 22, CCP22, FLJ16013, FLJ90719, POLYDOM, selectin-like protein, SELOB, SEL-OB, sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1, sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1 | |
| Synthetic peptide containing human SVEP1 Peptide Accession # Q4LDE5 | |
| 100 μg | |
| 79987 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 987420 | |
| Western Blot 2μg/mL | |
| Q4LDE5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human SVEP1 in direct Western blots. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction