missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SMURF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £357.00
Specifications
| Antigen | SMURF1 |
|---|---|
| Clone | 530131 |
| Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18760569
![]() |
R&D Systems
MAB9507-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706659
![]() |
R&D Systems
MAB9507-100 |
100 μg |
£357.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMURF1 Monoclonal specifically detects SMURF1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
| SMURF1 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| E3 ubiquitin ligase SMURF1, EC 6.3.2, EC 6.3.2.-, hSMURF1, KIAA1625, KIAA1625E3 ubiquitin-protein ligase SMURF1, SMAD specific E3 ubiquitin protein ligase 1, SMAD ubiquitination regulatory factor 1, Smad-specific E3 ubiquitin ligase 1, SMAD-specific E3 ubiquitin-protein ligase 1 | |
| E. coli-derived recombinant human SMURF1 Met496-Glu757 Accession # Q9HCE7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human SMURF1 in direct ELISAs. |
| 530131 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HCE7 | |
| 57154 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title