missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SMURF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9507-SP
This item is not returnable.
View return policy
Description
SMURF1 Monoclonal specifically detects SMURF1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| SMURF1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| E3 ubiquitin ligase SMURF1, EC 6.3.2, EC 6.3.2.-, hSMURF1, KIAA1625, KIAA1625E3 ubiquitin-protein ligase SMURF1, SMAD specific E3 ubiquitin protein ligase 1, SMAD ubiquitination regulatory factor 1, Smad-specific E3 ubiquitin ligase 1, SMAD-specific E3 ubiquitin-protein ligase 1 | |
| E. coli-derived recombinant human SMURF1 Met496-Glu757 Accession # Q9HCE7 | |
| 25 μg | |
| Primary | |
| Detects human SMURF1 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 530131 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Q9HCE7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57154 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction