missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SMPD1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £403.00
Specifications
| Antigen | SMPD1 |
|---|---|
| Clone | 563418 |
| Concentration | LYOPH |
| Dilution | Immunoprecipitation 25 ug/mL, Neutralization 0.384 ug/mL |
| Applications | Immunoprecipitation, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737079
![]() |
R&D Systems
MAB5348-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18160911
![]() |
R&D Systems
MAB5348 |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMPD1 Monoclonal specifically detects SMPD1 in Human samples. It is validated for Immunoprecipitation, Neutralization.Specifications
| SMPD1 | |
| LYOPH | |
| Immunoprecipitation, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_000534 | |
| 6609 | |
| Chinese hamster ovary cell line CHO-derived recombinant human SMPD1 isoform 1 His62-Pro628 Accession # NP_000534 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human SMPD1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human SMPD3 and recombinant mouse SMPD1 is observed. |
| 563418 | |
| Immunoprecipitation 25 ug/mL, Neutralization 0.384 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal | |
| SMPD1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title