missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SMPD1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5348-SP
This item is not returnable.
View return policy
Description
SMPD1 Monoclonal specifically detects SMPD1 in Human samples. It is validated for Immunoprecipitation, Neutralization.
Specifications
| SMPD1 | |
| Monoclonal | |
| LYOPH | |
| Immunoprecipitation 25 ug/mL, Neutralization 0.384 ug/mL | |
| NP_000534 | |
| SMPD1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human SMPD1 isoform 1 His62-Pro628 Accession # NP_000534 | |
| 25 μg | |
| Primary | |
| Detects human SMPD1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human SMPD3 and recombinant mouse SMPD1 is observed. | |
| Human | |
| Purified |
| Immunoprecipitation, Neutralization | |
| 563418 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6609 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction