missing translation for 'onlineSavingsMsg'
Learn More

Human SLC7A5/LAT1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£171.00 - £436.00

Specifications

Antigen SLC7A5/LAT1
Clone BU53
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18734347
GREENER_CHOICE
View Documents
R&D Systems
MAB10390-SP
25 μg
£171.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18763757
GREENER_CHOICE
View Documents
R&D Systems
MAB10390-100
100 μg
£436.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC7A5/LAT1 Monoclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

SLC7A5/LAT1
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
4F2 LC, 4F2LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1, LAT1hLAT1, MPE16, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, TA1, y+ system cationic amino acid transporter
Plasma cell line 8226, Accession # Q01650
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects Human SLC7A5/LAT1 in direct ELISAs.
BU53
Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
Q01650
8140
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.