missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC7A5/LAT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10390-100
This item is not returnable.
View return policy
Description
SLC7A5/LAT1 Monoclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| SLC7A5/LAT1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 4F2 LC, 4F2LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1, LAT1hLAT1, MPE16, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, TA1, y+ system cationic amino acid transporter | |
| Plasma cell line 8226, Accession # Q01650 | |
| 100 μg | |
| Primary | |
| Detects Human SLC7A5/LAT1 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| BU53 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q01650 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8140 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction