missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-7/CD328 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£206.00 - £532.00
Specifications
| Antigen | Siglec-7/CD328 |
|---|---|
| Clone | 194211 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719108
![]() |
R&D Systems
FAB11381A-025 |
25 Tests |
£206.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18114921
|
R&D Systems
FAB11381A-100 |
100 Tests |
£532.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Flow Cytometry.Specifications
| Siglec-7/CD328 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7 | |
| SIGLEC7 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Siglec-7 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-6, -8, -9 or recombinant Siglec-E is observed. |
| 194211 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| Q9Y286 | |
| 27036 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-7 Gln19-Gly357 (predicted) Accession # Q9Y286 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title