missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-7/CD328 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB11381A-100
This item is not returnable.
View return policy
Description
Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-7/CD328 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27036 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 194211 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9Y286 | |
| SIGLEC7 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-7 Gln19-Gly357 (predicted) Accession # Q9Y286 | |
| 100 Tests | |
| Primary | |
| Detects human Siglec-7 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Siglec-6, -8, -9 or recombinant Siglec-E is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction