missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin E2/PN1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £369.00
Specifications
| Antigen | Serpin E2/PN1 |
|---|---|
| Clone | 449888 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728829
![]() |
R&D Systems
MAB2980-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18100971
|
R&D Systems
MAB2980 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blot.Specifications
| Serpin E2/PN1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001130000 | |
| 5270 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin E2/PN1 Met1-Pro397 Accession # NP_001130000 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Serpin E2/PN1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A12, B6, B8, B9, C1, D1, F2, I1, recombinant mouse Serpin E2, or rhPEDF is observed. |
| 449888 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| SERPINE2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title