missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin E2/PN1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2980
This item is not returnable.
View return policy
Description
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blot.
Specifications
| Serpin E2/PN1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001130000 | |
| SERPINE2 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin E2/PN1 Met1-Pro397 Accession # NP_001130000 | |
| 100 μg | |
| Primary | |
| Detects human Serpin E2/PN1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A12, B6, B8, B9, C1, D1, F2, I1, recombinant mouse Serpin E2, or rhPEDF is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 449888 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5270 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction