missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin B8/Proteinase Inhibitor 8 Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £432.00
Specifications
| Antigen | Serpin B8/Proteinase Inhibitor 8 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789257
![]() |
R&D Systems
AF4158-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18002022
![]() |
R&D Systems
AF4158 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin B8/Proteinase Inhibitor 8 Polyclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CAP2, CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, NK10, Peptidase inhibitor 8, PI8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| SERPINB8 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P50452 | |
| 5271 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin B8/Proteinase Inhibitor 8 Asp2-Pro374 Accession # P50452 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Serpin B8/Proteinase Inhibitor 8 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Serpin B8 is observed and less than 2% cross-reactivity with recombinant human (rh) Serpin B6, rhSerpin B9, and rhSerpin A5 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title