missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin B8/Proteinase Inhibitor 8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4158
This item is not returnable.
View return policy
Description
Serpin B8/Proteinase Inhibitor 8 Polyclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| Polyclonal | |
| Unconjugated | |
| P50452 | |
| SERPINB8 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Serpin B8/Proteinase Inhibitor 8 Asp2-Pro374 Accession # P50452 | |
| 100 μg | |
| Primary | |
| Detects human Serpin B8/Proteinase Inhibitor 8 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Serpin B8 is observed and less than 2% cross-reactivity with recombinant human (rh) Serpin B6, rhSerpin B9, and rhSerpin A5 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| CAP2, CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, NK10, Peptidase inhibitor 8, PI8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5271 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction