missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 6C Antibody, R&D Systems™

Mouse Monoclonal Antibody
£147.00 - £452.00
Specifications
| Antigen | Semaphorin 6C |
|---|---|
| Clone | 254506 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743788
![]() |
R&D Systems
MAB2219-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18150211
![]() |
R&D Systems
MAB2219 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Semaphorin 6C Monoclonal specifically detects Semaphorin 6C in Human samples. It is validated for Western Blot.Specifications
| Semaphorin 6C | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY | |
| SEMA6C | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 254506 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_112175 | |
| 10500 | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6C Ala25-Val601 Accession # NP_112175 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Semaphorin 3A, 3B, 6A, 6B, 6D, 7A, recombinant mouse Semaphorin 3C, 3E, 3F, or 6C is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title