missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 6C Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2219
This item is not returnable.
View return policy
Description
Semaphorin 6C Monoclonal specifically detects Semaphorin 6C in Human samples. It is validated for Western Blot.
Specifications
| Semaphorin 6C | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10500 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 254506 | |
| Western Blot 1 ug/mL | |
| NP_112175 | |
| SEMA6C | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6C Ala25-Val601 Accession # NP_112175 | |
| 500 μg | |
| Primary | |
| Detects human Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Semaphorin 3A, 3B, 6A, 6B, 6D, 7A, recombinant mouse Semaphorin 3C, 3E, 3F, or 6C is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction