missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human S100A8/S100A9 Heterodimer Quantikine ELISA Kit
Description
The Human S100A8/S100A9 Heterodimer Quantikine ELISA Kit quantifies human S100A8/S100A9 Heterodimer in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (10 ul), urine (50 ul), fecal extract (50 ul), human milk (10 ul).
Specifications
Specifications
| Assay Range | 0.63 - 40ng/mL |
| Assay Sensitivity | 0.215ng/mL |
| Conjugate | HRP |
| Cross Reactivity | <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested. |
| Sample Type | Cell Culture Supernatant, Fecal Extract, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Tissue Lysates, Urine |
| Specificity | Natural and Recombinant Human S100A8/S100A9 Heterodimer. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (10uL),Urine (50uL),Fecal Extract (50uL),Human Milk (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?