missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR gamma t/RORC2/NR1F3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £436.00
Specifications
| Antigen | ROR gamma t/RORC2/NR1F3 |
|---|---|
| Clone | 1181A |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792878
![]() |
R&D Systems
MAB61093-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783328
![]() |
R&D Systems
MAB61093-100 |
100 μg |
£436.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| ROR gamma t/RORC2/NR1F3 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Rabbit | |
| Human | |
| P51449 | |
| E.coli derived recombinant human ROR gamma T/RORC2/NR1F3 Met1-Gln100 Accession # P51449 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human ROR gamma t/RORC2/NR1F3 by direct ELISAs. |
| 1181A | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 6097 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title