missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR gamma t/RORC2/NR1F3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB61093-100
This item is not returnable.
View return policy
Description
ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ROR gamma t/RORC2/NR1F3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6097 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry | |
| 1181A | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| P51449 | |
| E.coli derived recombinant human ROR gamma T/RORC2/NR1F3 Met1-Gln100 Accession # P51449 | |
| 100 μg | |
| Primary | |
| Detects recombinant human ROR gamma t/RORC2/NR1F3 by direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction