missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROBO3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
£180.00 - £511.00
Specifications
| Antigen | ROBO3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774757
![]() |
R&D Systems
AF3076-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18022062
![]() |
R&D Systems
AF3076 |
100 μg |
£511.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROBO3 Polyclonal specifically detects ROBO3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| ROBO3 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| FLJ21044, HGPPS, HGPS, RBIG1horizontal gaze palsy with progressive scoliosis, retinoblastoma inhibiting gene 1, RIG1, roundabout (axon guidance receptor, Drosophila) homolog 3, roundabout homolog 3, roundabout, axon guidance receptor, homolog 3 (Drosophila), Roundabout-like protein 3 | |
| ROBO3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q96MS0 | |
| 64221 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO3 Leu40-Ser545 Accession # Q96MS0 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects ROBO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse ROBO3 is observed, 5% cross-reactivity with recombinant human ROBO2 and recombinant rat ROBO1 is observed, and less than 1% cross-reactivity with recombinant human ROBO4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title