missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROBO3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF3076-SP
This item is not returnable.
View return policy
Description
ROBO3 Polyclonal specifically detects ROBO3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ROBO3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ21044, HGPPS, HGPS, RBIG1horizontal gaze palsy with progressive scoliosis, retinoblastoma inhibiting gene 1, RIG1, roundabout (axon guidance receptor, Drosophila) homolog 3, roundabout homolog 3, roundabout, axon guidance receptor, homolog 3 (Drosophila), Roundabout-like protein 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64221 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q96MS0 | |
| ROBO3 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO3 Leu40-Ser545 Accession # Q96MS0 | |
| 25 μg | |
| Primary | |
| Detects ROBO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse ROBO3 is observed, 5% cross-reactivity with recombinant human ROBO2 and recombinant rat ROBO1 is observed, and less than 1% cross-reactivity with recombinant human ROBO4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction