missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROBO3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF3076
This item is not returnable.
View return policy
Description
ROBO3 Polyclonal specifically detects ROBO3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ROBO3 | |
| Polyclonal | |
| Unconjugated | |
| Q96MS0 | |
| ROBO3 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO3 Leu40-Ser545 Accession # Q96MS0 | |
| 100 μg | |
| Primary | |
| Detects ROBO3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse ROBO3 is observed, 5% cross-reactivity with recombinant human ROBO2 and recombinant rat ROBO1 is observed, and less than 1% cross-reactivity with recombinant human ROBO4 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| FLJ21044, HGPPS, HGPS, RBIG1horizontal gaze palsy with progressive scoliosis, retinoblastoma inhibiting gene 1, RIG1, roundabout (axon guidance receptor, Drosophila) homolog 3, roundabout homolog 3, roundabout, axon guidance receptor, homolog 3 (Drosophila), Roundabout-like protein 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64221 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto