missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RGM-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £370.00
Specifications
| Antigen | RGM-B |
|---|---|
| Clone | 398528 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729138
![]() |
R&D Systems
MAB3630-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18118980
|
R&D Systems
MAB3630 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGM-B Monoclonal specifically detects RGM-B in Human samples. It is validated for Western Blot.Specifications
| RGM-B | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6NW40 | |
| 285704 | |
| Mouse myeloma cell line NS0-derived recombinant human RGM-B isoform 1 Gly46-Ser411 Accession # Q6NW40 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human RGM-B in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse RGM-B, recombinant human (rh) RGM-A, or rhRGM-C is observed. |
| 398528 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp434P228, Dragon, DRG11-responsive axonal guidance and outgrowth of neurite, RGM domain family, member B, RGMB | |
| RGMB | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title