missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RGM-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3630-SP
This item is not returnable.
View return policy
Description
RGM-B Monoclonal specifically detects RGM-B in Human samples. It is validated for Western Blot.
Specifications
| RGM-B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6NW40 | |
| RGMB | |
| Mouse myeloma cell line NS0-derived recombinant human RGM-B isoform 1 Gly46-Ser411 Accession # Q6NW40 | |
| 25 μg | |
| Primary | |
| Detects human RGM-B in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse RGM-B, recombinant human (rh) RGM-A, or rhRGM-C is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 398528 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp434P228, Dragon, DRG11-responsive axonal guidance and outgrowth of neurite, RGM domain family, member B, RGMB | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 285704 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction