missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein Rabbit anti-Virus, Clone: R338, Novus Biologicals™
Rabbit Monoclonal Antibody
£395.00 - £523.00
Specifications
| Antigen | Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein |
|---|---|
| Clone | R338 |
| Dilution | Sandwich ELISA Capture 0.5-4 ug/ml |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18216956
|
Novus Biologicals
NBP2-89761-200UG |
200 μg |
£395.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243876
|
Novus Biologicals
NBP2-89761-500UG |
500 μg |
£523.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein Monoclonal specifically detects Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein in Virus samples. It is validated for Sandwich ELISA Capture, Block/Neutralize.Specifications
| Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein | |
| Sandwich ELISA Capture 0.5-4 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| Recombinant Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| R338 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| HRSVgp08, RSV, RSV fusion protein | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title